Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

760315

Sigma-Aldrich

Imidazo[1,2-a]pyridine-5-carboxaldehyde

97%

Synonym(s):

5-Formylimidazo[1,2-a]pyridine, Imidazo[1,2-a]pyridine-5-carbaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6N2O
CAS Number:
Molecular Weight:
146.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

123-126 °C

functional group

aldehyde

SMILES string

O=Cc1cccc2nccn12

InChI

1S/C8H6N2O/c11-6-7-2-1-3-8-9-4-5-10(7)8/h1-6H

InChI key

IZZPLXLDWJCBDF-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service