Skip to Content
MilliporeSigma
All Photos(1)

Documents

76030

Sigma-Aldrich

Palladium on calcium carbonate

~10% Pd basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd
MDL number:
UNSPSC Code:
12141733
eCl@ss:
38191001
PubChem Substance ID:
NACRES:
NA.22

form

powder

autoignition temp.

>1120 °F

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

concentration

~10% Pd

SMILES string

[Ca++].[Pd++].[O-]C([O-])=O.[O-]C([O-])=O

InChI

1S/2CH2O3.Ca.Pd/c2*2-1(3)4;;/h2*(H2,2,3,4);;/q;;2*+2/p-4

InChI key

XGLHAVVVUOEGIO-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Aqueous Heck coupling and hydroarylation
  • Reduction reactions
  • Hydrogenation reactions
Hydrogenation catalyst

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service