Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

760293

Sigma-Aldrich

2,4,6,8-Tetramethylcyclotetrasiloxane

packaged for use in deposition systems

Synonym(s):

1,3,5,7-Cyclotetra(methylsiloxane), 1,3,5,7-Tetramethylcyclotetrasiloxane, TMCTS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HSiCH3O)4
CAS Number:
Molecular Weight:
240.51
Beilstein:
1775282
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

deposition grade

Assay

≥99.5% (GC)
≥99.999% (trace metals analysis)

form

liquid

refractive index

n20/D 1.387 (lit.)

bp

134 °C (lit.)

mp

−69 °C (lit.)

density

0.986 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[SiH]1O[SiH](C)O[SiH](C)O[SiH](C)O1

InChI

1S/C4H16O4Si4/c1-9-5-10(2)7-12(4)8-11(3)6-9/h9-12H,1-4H3

InChI key

BQYPERTZJDZBIR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2, 4, 6, 8-Tetramethylcyclotetrasiloxane (TMCTS) is an important precursor for the deposition of polysiloxane, cyclic siloxane, silicon dioxide, oxycarbide thin films with low dielectric constant for microelectronics, semiconductors and other applications

Features and Benefits

Precursor for gate dielectrics in thin-film transistors (TFT), and is a component of photochemically formed SiOX monolayers on TiO.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ward, L. J.; et al.
Langmuir, 19, 2110-2110 (2003)
Pai, C. S.; et al.
Materials Chemistry and Physics, 44, 1-1 (1996)
Dae-Yoon Kim et al.
Soft matter, 11(1), 58-68 (2014-11-20)
Unconventional star-shaped liquid crystals (abbreviated as SiLCs) were successfully synthesized by chemically connecting four cyanobiphenyl anisotropic mesogens to the periphery of a super-hydrophobic and ultra-flexible cyclic tetramethyltetrasiloxane ring with flexible hexyl chains. Based on the combined experimental techniques of differential

Questions

  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service