Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

76029

Sigma-Aldrich

3-(Trifluoromethyl)-D-phenylalanine

≥96.0% (HPLC)

Synonym(s):

(R)-2-Amino-3-[3-(trifluoromethyl)phenyl]propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10F3NO2
CAS Number:
Molecular Weight:
233.19
Beilstein:
5048167
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥96.0% (HPLC)

SMILES string

N[C@H](Cc1cccc(c1)C(F)(F)F)C(O)=O

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service