Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

759198

Sigma-Aldrich

Di(1-adamantyl)-1-piperidinylphenylphosphine

95%

Synonym(s):

2-(Di-1-adamantylphosphino)phenylpiperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H44NP
CAS Number:
Molecular Weight:
461.66
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

reaction suitability

reagent type: ligand

mp

225-230 °C

functional group

phosphine

SMILES string

[H][C@]1(C[C@](C2)([H])C3)C[C@]3([H])C[C@]2([P@@](C4=CC=CC=C4N5CCCCC5)[C@@]67C[C@@](C8)([H])C[C@@](C[C@@]8([H])C7)([H])C6)C1

InChI

1S/C31H44NP/c1-4-8-32(9-5-1)28-6-2-3-7-29(28)33(30-16-22-10-23(17-30)12-24(11-22)18-30)31-19-25-13-26(20-31)15-27(14-25)21-31/h2-3,6-7,22-27H,1,4-5,8-21H2/t22-,23+,24-,25-,26+,27-,30-,31-

InChI key

FPQYIRBFZLNVLL-OSESXBJYSA-N

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DalPhos ligands enable Pd-catalyzed C-N and C-C bond formation with good functional group tolerance.

Related Content

Stradiotto group focuses on electronically rich ligands for late metal catalysis, including Pd and Au chemistry applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service