Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

758191

Sigma-Aldrich

3-Bromo-6-methoxy-2-methylpyridine

95%

Synonym(s):

3-Bromo-6-methoxy-2-picoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8BrNO
CAS Number:
Molecular Weight:
202.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.548

density

1.468 g/mL at 25 °C

functional group

bromo

storage temp.

2-8°C

SMILES string

COc1ccc(Br)c(C)n1

InChI

1S/C7H8BrNO/c1-5-6(8)3-4-7(9-5)10-2/h3-4H,1-2H3

InChI key

VWNXCCTWVAQAPL-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

222.8 °F

Flash Point(C)

106 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service