Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

756938

Sigma-Aldrich

Suberic-d12 acid dihydrazide

98 atom % D, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2NHCOCD2(CD2)4CD2CONHNH2
Molecular Weight:
214.33
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

mp

140-144 °C (lit.)

mass shift

M+12

SMILES string

[2H]C([2H])(C(=O)NN)C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)NN

InChI

1S/C8H18N4O2/c9-11-7(13)5-3-1-2-4-6-8(14)12-10/h1-6,9-10H2,(H,11,13)(H,12,14)/i1D2,2D2,3D2,4D2,5D2,6D2

InChI key

HATIEXJZXOLRAO-LBTWDOQPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

410.0 °F - closed cup

Flash Point(C)

210 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service