Skip to Content
MilliporeSigma
All Photos(1)

Documents

756644

Sigma-Aldrich

N,N,N′,N′- Tetramethyl-1,3-phenylenediamine

97%

Synonym(s):

1,3-Bis(dimethylamino)benzene, N,N,N′,N′-Tetramethyl-m-phenylenediamine, N1,N1,N3,N3-Tetramethylbenzene-1,3-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2
CAS Number:
Molecular Weight:
164.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

granular

refractive index

n20/D 1.580

density

0.976 g/mL at 25 °C

SMILES string

CN(C)c1cccc(c1)N(C)C

InChI

1S/C10H16N2/c1-11(2)9-6-5-7-10(8-9)12(3)4/h5-8H,1-4H3

InChI key

LFZLVJBOEONQHV-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service