Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

756547

Sigma-Aldrich

Strontium silicate aluminate, europium and dysprosium doped

97% trace metals basis

Synonym(s):

Europium and dysprosium doped strontium aluminum silicon oxide, Long persistent blue phosphor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sr2.90Eu0.03Dy0.07Al4SiO11
Molecular Weight:
582.04
UNSPSC Code:
12352300

description

long persistent blue phosphor

Assay

97% trace metals basis

form

powder

composition

Sr2.90Eu0.03Dy0.07Al4SiO11

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

particle size

-300 mesh

mp

>1200 °C (lit.)

fluorescence

λem Blue (UV excitation source)

General description

Long persistent blue phosphor

Other Notes

Persist Time (min): 3500 ± 5%

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiu, Z.; Zhou, Y.; Lu, M.; Zhang, A.; Ma, Q.
Solid State Sciences, 10, 629-629 (2008)
Mishra, S.B.; et al.
Journal of Applied Polymer Science. Applied Polymer Symposium, 115, 579-579 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service