Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

756253

Sigma-Aldrich

16-Bromo-1-hexadecanol

Synonym(s):

1-Bromohexadecane-16-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H33BrO
CAS Number:
Molecular Weight:
321.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

45-52 °C

SMILES string

OCCCCCCCCCCCCCCCCBr

InChI

1S/C16H33BrO/c17-15-13-11-9-7-5-3-1-2-4-6-8-10-12-14-16-18/h18H,1-16H2

InChI key

GOSQZSJMSMTIFI-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service