Skip to Content
MilliporeSigma
All Photos(1)

Documents

756180

Sigma-Aldrich

3,5-Dimethoxytriphenylamine

97%

Synonym(s):

3,5-Dimethoxy-N,N-diphenylaniline, 5-(Diphenylamino)-1,3-dimethoxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19NO2
Molecular Weight:
305.37
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

98-103 °C

SMILES string

COc1cc(OC)cc(c1)N(c2ccccc2)c3ccccc3

InChI

1S/C20H19NO2/c1-22-19-13-18(14-20(15-19)23-2)21(16-9-5-3-6-10-16)17-11-7-4-8-12-17/h3-15H,1-2H3

InChI key

HNNBPHRCCXLGQP-UHFFFAOYSA-N

Application

Novel triphenylamine derivative

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service