Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

75604

Sigma-Aldrich

Trimethyl orthopropionate

≥95.0% (T)

Synonym(s):

1,1,1-Trimethoxypropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2C(OCH3)3
CAS Number:
Molecular Weight:
134.17
Beilstein:
1736043
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (T)

form

liquid

refractive index

n20/D 1.398

bp

121-122 °C (lit.)

density

0.944 g/mL at 20 °C (lit.)

functional group

ether

SMILES string

CCC(OC)(OC)OC

InChI

1S/C6H14O3/c1-5-6(7-2,8-3)9-4/h5H2,1-4H3

InChI key

ZGMNAIODRDOMEK-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

66.2 °F - closed cup

Flash Point(C)

19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jessica Granados-Pineda et al.
Molecules (Basel, Switzerland), 23(4) (2018-04-13)
Propolis is a resinous beehive product that has been used worldwide in traditional medicine to prevent and treat colds, wounds, rheumatism, heart disease and diabetes. Diabetic nephropathy is the final stage of renal complications caused by diabetes and for its
Lakshmi Priya Pasari et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 112, 108629-108629 (2019-02-25)
Acute pancreatitis (AP) is an exocrine dysfunction of the pancreas where oxidative stress and inflammatory cytokines play a key role in induction and progression of the disease. Studies have demonstrated that antioxidant phytochemicals have been effective in improving pancreatitis condition

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service