Skip to Content
MilliporeSigma
All Photos(1)

Documents

756024

Sigma-Aldrich

1-Naphthyl methacrylate

contains <500 ppm monomethyl ether hydroquinone (as inhibitor), 97% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12O2
CAS Number:
Molecular Weight:
212.24
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97% (GC)

form

liquid

contains

<500 ppm monomethyl ether hydroquinone (as inhibitor)

refractive index

n20/D 1.588

density

1.149 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)Oc1cccc2ccccc12

InChI

1S/C14H12O2/c1-10(2)14(15)16-13-9-5-7-11-6-3-4-8-12(11)13/h3-9H,1H2,2H3

InChI key

HVYCQBKSRWZZGX-UHFFFAOYSA-N

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service