Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

75578

Sigma-Aldrich

4-Amino-1-Boc-piperidine hydrochloride

≥97.0% (N)

Synonym(s):

tert-Butyl 4-amino-1-piperidinecarboxylate hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20N2O2 · HCl
CAS Number:
Molecular Weight:
236.74
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.0% (N)

form

powder

SMILES string

Cl[H].CC(C)(C)OC(=O)N1CCC(N)CC1

InChI

1S/C10H20N2O2.ClH/c1-10(2,3)14-9(13)12-6-4-8(11)5-7-12;/h8H,4-7,11H2,1-3H3;1H

InChI key

KLOQZAJAQQKCNC-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Mono-Boc-protected diamines are versatile building blocks for chemical synthesis. Their production is a lot more challenging than the simple reaction scheme might imply, because the Boc-anhydride reagent cannot differentiate between the two identical amino moieties in the substrate.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service