Skip to Content
MilliporeSigma
All Photos(1)

Documents

755621

Sigma-Aldrich

Glyceryl 1,2-di(oleate-13C18) 3-oleate

≥99% 13C, ≥97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C36C21H104O6
Molecular Weight:
921.17
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

isotopic purity

≥99% 13C

Assay

≥97% (CP)

form

liquid

technique(s)

bio NMR: suitable

mass shift

M+36

storage temp.

−20°C

SMILES string

O=C(CCCCCCC/C=C\CCCCCCCC)OCC(O[13C]([13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2]/[13CH]=[13CH]\[13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH3])=O)CO[13C]([13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2]/[13CH]=[13CH]\[13CH2][13CH2][13CH2][1

InChI

1S/C57H104O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h25-30,54H,4-24,31-53H2,1-3H3/b28-25-,29-26-,30-27-/i1+1,3+1,4+1,6+1,7+1,9+1,10+1,12+1,13+1,15+1,16+1,18+1,19+1,21+1,22+1,24+1,25+1,27+1,28+1,30+1,31+1,33+1,34+1,36+1,37+1,39+1,40+1,42+1,43+1,45+1,46+1,48+1,49+1,51+1,55+1,57+1

InChI key

PHYFQTYBJUILEZ-FAHXWPFJSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids not in Storage Class 3

WGK

awg

Flash Point(F)

626.0 °F - closed cup

Flash Point(C)

330.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service