Skip to Content
MilliporeSigma
All Photos(1)

Documents

755540

Sigma-Aldrich

Ethyl 2,2-dibromodiethylphosphonoacetate

97%

Synonym(s):

2,2-Dibromo-2-(diethoxyphosphinyl)acetic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15Br2O5P
CAS Number:
Molecular Weight:
381.98
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

Assay

97%

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.493

density

1.636 at 25 °C

SMILES string

CCOC(=O)C(Br)(Br)P(=O)(OCC)OCC

InChI

1S/C8H15Br2O5P/c1-4-13-7(11)8(9,10)16(12,14-5-2)15-6-3/h4-6H2,1-3H3

InChI key

XKZDOIGWQFLTMZ-UHFFFAOYSA-N

Application

Coupling partner in Suzuki-Miyuara C-C bond formations

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service