Skip to Content
MilliporeSigma
All Photos(1)

Documents

755532

Sigma-Aldrich

Butyraldehyde-d8

≥99 atom % D, ≥98% (CP)

Synonym(s):

Butanal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CD2CD2CDO
Molecular Weight:
80.16
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

isotopic purity

≥99 atom % D

Assay

≥98% (CP)

bp

75 °C (lit.)

mp

-96 °C (lit.)

density

0.887 g/mL at 25 °C

mass shift

M+8

SMILES string

[2H]C(C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=O

InChI

1S/C4H8O/c1-2-3-4-5/h4H,2-3H2,1H3/i1D3,2D2,3D2,4D

InChI key

ZTQSAGDEMFDKMZ-RIZALVEQSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

23.0 °F

Flash Point(C)

-5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service