Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

755273

Sigma-Aldrich

2-Chloro-5-fluorobenzylamine

95%

Synonym(s):

(2-Chloro-5-fluorophenyl)methanamine, 2-Chloro-5-fluorobenzenemethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7ClFN
CAS Number:
Molecular Weight:
159.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.537

density

1.283 g/mL at 25 °C

functional group

amine
chloro
fluoro

SMILES string

NCc1cc(F)ccc1Cl

InChI

1S/C7H7ClFN/c8-7-2-1-6(9)3-5(7)4-10/h1-3H,4,10H2

InChI key

IGFBMQSUHFQVFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

226.4 °F

Flash Point(C)

108 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service