754781
3-Ethylcarbazole
97%
Synonym(s):
3-Ethyl-9H-carbazole
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C14H13N
CAS Number:
Molecular Weight:
195.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
148-152 °C
SMILES string
CCc1ccc2[nH]c3ccccc3c2c1
InChI
1S/C14H13N/c1-2-10-7-8-14-12(9-10)11-5-3-4-6-13(11)15-14/h3-9,15H,2H2,1H3
InChI key
DJMRXHOKZAERIV-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service