Skip to Content
MilliporeSigma
All Photos(1)

Documents

754277

Sigma-Aldrich

(3-Glycidyl-13C3-oxypropyl) trimethoxysilane

99 atom % 13C, 97% (CP)

Synonym(s):

3-(2,3-Epoxypropoxy)propyltrimethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3C6H20O5Si
Molecular Weight:
239.32
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

97% (CP)

form

liquid

bp

120 °C/2 mmHg (lit.)

density

1.083 g/mL at 25 °C

mass shift

M+3

SMILES string

CO[Si](OC)(OC)CCCO[13CH2][13CH]1O[13CH2]1

InChI

1S/C9H20O5Si/c1-10-15(11-2,12-3)6-4-5-13-7-9-8-14-9/h9H,4-8H2,1-3H3/i7+1,8+1,9+1

InChI key

BPSIOYPQMFLKFR-ULEDQSHZSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

275.0 °F - closed cup

Flash Point(C)

135 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service