Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

754064

Sigma-Aldrich

DH-FTTF

sublimed, 95%

Synonym(s):

5,5′-Bis(7-hexyl-9H-fluoren-2-yl)-2,2′-bithiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H46S2
CAS Number:
Molecular Weight:
662.99
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

95%

form

sublimed

loss

0.5 wt. %, 405 °C

mp

373-378 °C

semiconductor properties

P-type (mobility=0.05-0.12 cm2/V·s)

InChI

1S/C46H46S2/c1-3-5-7-9-11-31-13-17-39-35(25-31)29-37-27-33(15-19-41(37)39)43-21-23-45(47-43)46-24-22-44(48-46)34-16-20-42-38(28-34)30-36-26-32(14-18-40(36)42)12-10-8-6-4-2/h13-28H,3-12,29-30H2,1-2H3

InChI key

IEOMVXSYYUOEQW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Robust small molecule p-channel semiconductor that exhibits excellent device performance and resistant to harsh environments including aqueous conditions for BioMEMS devices.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic thin-film transistors fabricated on resorbable biomaterial substrates.
Christopher J Bettinger et al.
Advanced materials (Deerfield Beach, Fla.), 22(5), 651-655 (2010-03-11)

Articles

Materials Design Concepts for Efficient Blue OLEDs: A Joint Theoretical and Experimental Study

Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.

Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service