Skip to Content
MilliporeSigma
All Photos(1)

Documents

753912

Sigma-Aldrich

SMDPPEH

sublimed, 97% (HPLC)

Synonym(s):

2,5-Di-(2-ethylhexyl)-3,6-bis-(5′′-n-hexyl-[2,2′,5′,2′′]terthiophen-5-yl)-pyrrolo[3,4-c]pyrrole-1,4-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C58H72N2O2S6
CAS Number:
Molecular Weight:
1021.59
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

97% (HPLC)

form

sublimed

loss

0.5 wt. % TGA, 370 °C

λmax

644 nm in methylene chloride

Orbital energy

HOMO 5.2 eV 
LUMO 3.7 eV 

OPV Device Performance

ITO/PEDOT:PSS/SMDPPEH:PC71BM (1:1)/Al

  • Short-circuit current density (Jsc): 9.2 mA/cm2
  • Open-circuit voltage (Voc): 0.75 V
  • Fill Factor (FF): 0.44
  • Power Conversion Efficiency (PCE): 3 %

ITO/PEDOT:PSS/SMDPPEH:PC71BM (3:7)/Al
  • Short-circuit current density (Jsc): 7.7 mA/cm2
  • Open-circuit voltage (Voc): 0.72 V
  • Fill Factor (FF): 0.41
  • Power Conversion Efficiency (PCE): 2.4 %

ITO/PEDOT:PSS/SMDPPEH:PC71BM (7:3)/Al
  • Short-circuit current density (Jsc): 8.7 mA/cm2
  • Open-circuit voltage (Voc): 0.72 V
  • Fill Factor (FF): 0.35
  • Power Conversion Efficiency (PCE): 2.2 %

semiconductor properties

P-type (mobility~10−4 cm2/V·s)

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A low band gap, solution processable oligothiophene with a dialkylated diketopyrrolopyrrole chromophore for use in bulk heterojunction solar cells
Tamayo, A.; Dang, X.; Walker, B.; et al.
Applied Physics Letters, 94, 103301-103301 (2009)

Articles

Organic materials in optoelectronic devices like LEDs and solar cells are of significant academic and commercial interest.

Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.

Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service