Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

753440

Sigma-Aldrich

1-(3-Fluorophenyl)vinylboronic acid pinacol ester

97%

Synonym(s):

2-[1-(3-Fluorophenyl)vinyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18BFO2
CAS Number:
Molecular Weight:
248.10
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.499

density

1.041 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC1(C)OB(OC1(C)C)C(=C)c2cccc(F)c2

InChI

1S/C14H18BFO2/c1-10(11-7-6-8-12(16)9-11)15-17-13(2,3)14(4,5)18-15/h6-9H,1H2,2-5H3

InChI key

JHYKXYAWAWZTCM-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service