Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

753408

Sigma-Aldrich

5-Bromopyridine-2-acetic acid

97%

Synonym(s):

2-(5-Bromopyridin-2-yl)acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6BrNO2
CAS Number:
Molecular Weight:
216.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

125-130 °C

functional group

bromo
carboxylic acid

SMILES string

OC(=O)Cc1ccc(Br)cn1

InChI

1S/C7H6BrNO2/c8-5-1-2-6(9-4-5)3-7(10)11/h1-2,4H,3H2,(H,10,11)

InChI key

ATKULCGQSLCGEK-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service