Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

752754

Sigma-Aldrich

Tetraisopropyl vinylidenediphosphonate

technical grade, 70%

Synonym(s):

VDPE, Vinylidene-1,1-diphosphonic acid tetraisopropyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H30O6P2
CAS Number:
Molecular Weight:
356.33
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

70%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

SMILES string

CC(C)OP(=O)(OC(C)C)C(=C)P(=O)(OC(C)C)OC(C)C

InChI

1S/C14H30O6P2/c1-10(2)17-21(15,18-11(3)4)14(9)22(16,19-12(5)6)20-13(7)8/h10-13H,9H2,1-8H3

InChI key

IPUAADANTWKVJD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service