Skip to Content
MilliporeSigma
All Photos(4)

Documents

752576

Sigma-Aldrich

Fumaric acid-1,4-13C2,2,3-d2

greener alternative

≥99 atom % 13C, ≥98 atom % D, ≥98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOO13CCD=CD13COOH
CAS Number:
Molecular Weight:
120.07
UNSPSC Code:
12352106

isotopic purity

≥99 atom % 13C
≥98 atom % D

Assay

≥98% (CP)

form

solid

greener alternative product score

old score: 59
new score: 48
Find out more about DOZN™ Scoring

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Safer Solvents and Auxiliaries
Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

mass spectrometry (MS): suitable

mp

298-300 °C (subl.) (lit.)

greener alternative category

mass shift

M+4

SMILES string

O[13C](/C([2H])=C([2H])/[13C](O)=O)=O

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses”, “Safer Solvents and Auxiliaries” and “Inherently Safer Chemistry for Accident Prevention”. Click here to view its DOZN scorecard.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service