Skip to Content
MilliporeSigma
All Photos(1)

Documents

751812

Sigma-Aldrich

Methyl 2-chloro-4-iodopyridine-3-carboxylate

97%

Synonym(s):

2-Chloro-4-iodo-nicotinic acid methyl ester, Methyl 2-chloro-iodonicotinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5ClINO2
CAS Number:
Molecular Weight:
297.48
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

81-85 °C

SMILES string

COC(=O)c1c(Cl)nccc1I

InChI

1S/C7H5ClINO2/c1-12-7(11)5-4(9)2-3-10-6(5)8/h2-3H,1H3

InChI key

FXGPTKSMXUWJGO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service