Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

751383

Sigma-Aldrich

4′-Bromo-4-nitrostilbene

97%

Synonym(s):

1-Bromo-4-[2-(4-nitrophenyl)ethenyl]benzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10BrNO2
CAS Number:
Molecular Weight:
304.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

195-200 °C

SMILES string

[O-][N+](=O)c1ccc(\C=C\c2ccc(Br)cc2)cc1

InChI

1S/C14H10BrNO2/c15-13-7-3-11(4-8-13)1-2-12-5-9-14(10-6-12)16(17)18/h1-10H/b2-1+

InChI key

DFAOJKUHONBIMN-OWOJBTEDSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service