Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

750549

Sigma-Aldrich

6-Bromo-3-isopropyl-[1,2,4]triazolo[4,3-a]pyridine hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10BrN3 · HCl
CAS Number:
Molecular Weight:
276.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

241-246 °C

functional group

bromo

SMILES string

Cl.CC(C)c1nnc2ccc(Br)cn12

InChI

1S/C9H10BrN3.ClH/c1-6(2)9-12-11-8-4-3-7(10)5-13(8)9;/h3-6H,1-2H3;1H

InChI key

YIMXIQLCXJYRGO-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service