Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

749494

Sigma-Aldrich

(2-Ammonioethyl)di-tert-butylphosphonium bis(tetrafluoroborate)

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H26B2F8NP
Molecular Weight:
364.90
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

318-325 °C

functional group

phosphine

SMILES string

F[B-](F)(F)F.F[B-](F)(F)F.CC(C)(C)[PH+](CC[NH3+])C(C)(C)C

InChI

1S/C10H24NP.2BF4/c1-9(2,3)12(8-7-11)10(4,5)6;2*2-1(3,4)5/h7-8,11H2,1-6H3;;/q;2*-1/p+2

InChI key

MWVDZPSIELCQBF-UHFFFAOYSA-P

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service