Skip to Content
MilliporeSigma
All Photos(1)

Documents

748374

Sigma-Aldrich

Tetraphenylgermane

96%

Synonym(s):

Tetraphenyl germanium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ge(C6H5)4
CAS Number:
Molecular Weight:
381.06
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

reaction suitability

core: germanium
reagent type: catalyst

mp

230-235 °C (lit.)

SMILES string

c1ccc(cc1)[Ge](c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C24H20Ge/c1-5-13-21(14-6-1)25(22-15-7-2-8-16-22,23-17-9-3-10-18-23)24-19-11-4-12-20-24/h1-20H

InChI key

ILEXMONMGUVLRM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S D Warner et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 56(3), 453-466 (2000-05-04)
The effect of high external pressures on the vibrational spectra of the tetraphenyl Group 14 compounds, Ph4M (M = Si, Ge, Sn, Pb), were examined between ambient pressure and 40 kbar with the aid of a diamond-anvil cell. The four
Toan Trong Tran et al.
Science advances, 5(5), eaav9180-eaav9180 (2019-05-07)
Color centers in solids are the fundamental constituents of a plethora of applications such as lasers, light-emitting diodes, and sensors, as well as the foundation of advanced quantum information and communication technologies. Their photoluminescence properties are usually studied under Stokes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service