Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

748110

Sigma-Aldrich

SF7-AM

95% (HPLC)

Synonym(s):

SF7-AM, Bis(acetoxymethyl)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H23N7O12
CAS Number:
Molecular Weight:
685.55
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

95% (HPLC)

mp

160-165 °C

storage temp.

−20°C

SMILES string

CC(OCOC(CN(CC(OCOC(C)=O)=O)C(C1=CC=C(C(OC23C4=C(C=C(N=[N+]=[N-])C=C4)OC5C2C=CC(N=[N+]=[N-])=C5)=O)C3=C1)=O)=O)=O

InChI

1S/C31H25N7O12/c1-16(39)45-14-47-27(41)12-38(13-28(42)48-15-46-17(2)40)29(43)18-3-6-21-24(9-18)31(50-30(21)44)22-7-4-19(34-36-32)10-25(22)49-26-11-20(35-37-33)5-8-23(26)31/h3-11,22,25H,12-15H2,1-2H3

InChI key

KVUJKFQKNGAJST-UHFFFAOYSA-N

Application

SF7-AM is a fluorogenic; or "caged" rhodamine 110 derivative for the detection of hydrogen sulfide (H2S). Upon exposure to H2S; the azide functional groups are reduced to primary amines; restoring the fluorescence of the "caged" fluorophore. A fluorescence intensity increase of 20x is observed upon 1h of SF7-AM exposure to H2S. This activation of SF7-AM has been shown to detect H2S levels as low as 500 nM in vitro. SF7-AM has also been engineered specifically for use in live cell imaging of H2S through the addition of the acetoxymethyl ester groups; which are cleaved in vivo by esterases. This results in adding a net -2 charge to the probe; which was demonstrated to "trap" the probe in cells; thus increasing the sensitivity of the probe even when the cell culture media was changed.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cell-trappable fluorescent probes for endogenous hydrogen sulfide signaling and imaging H2O2-dependent H2S production.
Lin Vs, et al.
Proceedings of the National Academy of Sciences of the USA, 110(18), 7131?7135-7131?7135 null
Hang Zou et al.
Molecular plant-microbe interactions : MPMI, 32(8), 972-985 (2019-06-18)
The rhizobium-legume symbiotic system is crucial for nitrogen cycle balance in agriculture. Hydrogen sulfide (H2S), a gaseous signaling molecule, may regulate various physiological processes in plants. However, whether H2S has regulatory effect in this symbiotic system remains unknown. Herein, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service