Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

748048

Sigma-Aldrich

Methyl 2,3,4-tri-O-benzyl-6-O-trityl-α-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H46O6
CAS Number:
Molecular Weight:
706.86
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

solid

optical activity

[α]22/D +14.0, c = 0.5% in chloroform

mp

>300 °C

storage temp.

−20°C

SMILES string

CO[C@@H]1[C@H](OCC2=CC=CC=C2)[C@@H](OCC3=CC=CC=C3)[C@H](OCC4=CC=CC=C4)[C@@H](COC(C5=CC=CC=C5)(C6=CC=CC=C6)C7=CC=CC=C7)O1

InChI

1S/C47H46O6/c1-48-46-45(51-34-38-24-12-4-13-25-38)44(50-33-37-22-10-3-11-23-37)43(49-32-36-20-8-2-9-21-36)42(53-46)35-52-47(39-26-14-5-15-27-39,40-28-16-6-17-29-40)41-30-18-7-19-31-41/h2-31,42-46H,32-35H2,1H3/t42-,43-,44+,45-,46+/m1/s1

InChI key

AYWIUINDNNLVIG-PSKPMRIASA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service