Skip to Content
MilliporeSigma
All Photos(2)

Documents

747610

Sigma-Aldrich

9-Mesityl-10-methylacridinium perchlorate

95%

Synonym(s):

10-Methyl-9-(2,4,6-trimethylphenyl)acridinium perchlorate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22ClNO4
CAS Number:
Molecular Weight:
411.88
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

reaction suitability

core: acridinium
reaction type: Photocatalysis
reagent type: catalyst

photocatalyst activation

450 nm

SMILES string

CC1=CC(C)=CC(C)=C1C2=C3C(C=CC=C3)=[N+](C)C4=CC=CC=C42.O=Cl(=O)([O-])=O

InChI

1S/C23H22N.ClHO4/c1-15-13-16(2)22(17(3)14-15)23-18-9-5-7-11-20(18)24(4)21-12-8-6-10-19(21)23;2-1(3,4)5/h5-14H,1-4H3;(H,2,3,4,5)/q+1;/p-1

InChI key

LFMBERYWDLWXNO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service