Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74730

Sigma-Aldrich

Ocimene

analytical standard

Synonym(s):

3,7-Dimethyl-1,3,6-octatrien, 3,7-Dimethyl-1,3,6-octatriene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein:
1736172
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

analytical standard

Assay

≥90% (mixture of cis-ocimene and limonene, GC)

quality

reference standard

composition

limonene, 20-25%
cis-ocimene, 70-75%

refractive index

n20/D 1.485

bp

65-66 °C/13 mmHg

density

0.818 g/mL at 20 °C (lit.)

SMILES string

[H]C(CC([H])=C(C)C=C)=C(C)C

InChI

1S/C10H16/c1-5-10(4)8-6-7-9(2)3/h5,7-8H,1,6H2,2-4H3

InChI key

IHPKGUQCSIINRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefan Schulz et al.
Journal of chemical ecology, 34(1), 82-93 (2007-12-15)
Gilbert (1976) suggested that male-contributed odors of mated females of Heliconius erato could enforce monogamy. We investigated the pheromone system of a relative, Heliconius melpomene, using chemical analysis, behavioral experiments, and feeding experiments with labeled biosynthetic pheromone precursors. The abdominal
Jarmo K Holopainen et al.
Journal of chemical ecology, 36(10), 1068-1075 (2010-09-15)
Deciduous trees remobilize the nitrogen in leaves during the process of autumn coloration, thus providing a high quality food source for aphids preparing to lay over-wintering eggs. It has been suggested that aphids may use volatile organic compounds (VOCs) to:
Atsushi Muroi et al.
PloS one, 6(10), e24594-e24594 (2011-10-25)
A blend of volatile organic compounds (VOCs) emitted from plants induced by herbivory enables the priming of defensive responses in neighboring plants. These effects may provide insights useful for pest control achieved with transgenic-plant-emitted volatiles. We therefore investigated, under both
Takeshi Shimoda et al.
The New phytologist, 193(4), 1009-1021 (2012-01-17)
Plants under herbivore attack emit mixtures of volatiles (herbivore-induced plant volatiles, HIPVs) that can attract predators of the herbivores. Although the composition of HIPVs should be critical for the attraction, most studies of transgenic plant-emitted volatiles have simply addressed the
Fitsum Fikru Dube et al.
Malaria journal, 10, 375-375 (2011-12-21)
In the search for plant-based mosquito repellents, volatile emanations were investigated from five plant species, Corymbia citriodora, Ocimum suave, Ocimum lamiifolium, Olea europaea and Ostostegia integrifolia, traditionally used in Ethiopia as protection against mosquitoes. The behaviour of two mosquitoes, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service