Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

747297

Sigma-Aldrich

Oxalacetic acid-d4

97 atom % D, 95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DOOCCD2COCOOD
Molecular Weight:
136.10
UNSPSC Code:
12352115
PubChem Substance ID:

isotopic purity

97 atom % D

Assay

95% (CP)

form

solid

mass shift

M+4

storage temp.

−20°C

SMILES string

O=C(C(C([2H])([2H])C(O[2H])=O)=O)O[2H]

InChI

1S/C4H4O5/c5-2(4(8)9)1-3(6)7/h1H2,(H,6,7)(H,8,9)/i1D2/hD2

InChI key

KHPXUQMNIQBQEV-BGOGGDMHSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service