Skip to Content
MilliporeSigma
All Photos(1)

Documents

746703

Sigma-Aldrich

16-Mercaptohexadecanamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H33NOS
CAS Number:
Molecular Weight:
287.50
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder

mp

96-102 °C

storage temp.

2-8°C

SMILES string

O=C(CCCCCCCCCCCCCCCS)N

InChI

1S/C16H33NOS/c17-16(18)14-12-10-8-6-4-2-1-3-5-7-9-11-13-15-19/h19H,1-15H2,(H2,17,18)

InChI key

PIJCUJXBPYCINA-UHFFFAOYSA-N

Application

Amido derivative SAMs have been used to control the protein absorption in biological applications. “Wave function engineering” through charge transport by tunneling across self-assembled monolayers (SAMs) for molecular electronics.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"w-Terminated alkanethiolate monolayers on surfaces of copper; silver; and gold have similar wettabilities"
Laibinis; Paul E.; Whitesides; George M.
Journal of the American Chemical Society, 114, 1990-1990 (1992)
"Evaluation of amine- and amide-terminated self-assembled monolayers as "Molecular glues" for Au and SiO2 substrates"
Hooper; A. E.; Werho; D.; Hopson; T.; Palmer; O.
Surface and Interface Analysis : SIA, 31, 809-809 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service