Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

746630

Sigma-Aldrich

4,7-Dibromo-5-fluoro-2,1,3-benzothiadiazole

99% (HPLC)

Synonym(s):

Dibromo-FBT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6HBr2FN2S
CAS Number:
Molecular Weight:
311.96
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99% (HPLC)

form

solid

mp

158-163 °C

SMILES string

BrC1=C(F)C=C(Br)C2=NSN=C21

InChI

1S/C6HBr2FN2S/c7-2-1-3(9)4(8)6-5(2)10-12-11-6/h1H

InChI key

KVZDYOVYIHJETJ-UHFFFAOYSA-N

Application

Monomer for synthesis of polymers for high power conversion efficiency (PCE) organic solar cells and high mobility OFETs.
Fluorination lowers the polymer HOMO level; resulting in high open-circuit voltages well exceeding 0.7 V.
High-Efficiency Polymer F-PCPDTBT:PC70BM Bulk Heterojuction Organic Solar Cells (OPVs):
  • JSC = 14.1 mA/cm2
  • VOC = 0.74 V
  • FF = 0.58
  • PCE = 6.2%

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steve Albrecht et al.
Journal of the American Chemical Society, 134(36), 14932-14944 (2012-08-07)
A novel fluorinated copolymer (F-PCPDTBT) is introduced and shown to exhibit significantly higher power conversion efficiency in bulk heterojunction solar cells with PC(70)BM compared to the well-known low-band-gap polymer PCPDTBT. Fluorination lowers the polymer HOMO level, resulting in high open-circuit
Yong Zhang et al.
Chemical communications (Cambridge, England), 47(39), 11026-11028 (2011-09-13)
Small band-gap conjugated polymers based on monofluoro- and difluoro-substituted benzothiadiazole were developed. Highly efficient polymer solar cells (PCE as high as 5.40%) could be achieved for devices made from these polymers.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service