Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

745553

Sigma-Aldrich

Cobalt nanowires

powder, diam. × L 200-300 nm × 100-200 μm

Synonym(s):

Cobalt (nanoparticles), Cobalt element, Cobalt-59

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Co
CAS Number:
Molecular Weight:
58.93
EC Number:
UNSPSC Code:
12352302
NACRES:
NA.23

form

nanowires
powder

reaction suitability

reagent type: catalyst
core: cobalt

diam. × L

200-300 nm × 100-200 μm

storage temp.

2-8°C

InChI

1S/Co

InChI key

GUTLYIVDDKVIGB-UHFFFAOYSA-N

Application

Cobalt nanowires have been studied for their directional magnetic properties. In addition, cobalt nanowires have been studied for their magnetoresistance behavior at both room temperature and low temperatures. The metal nanostructure may be used to design thin film magnets with high energy density and high thermal stability.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1B - Skin Sens. 1 - STOT RE 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Low-temperature study of the magnetization reversal and magnetic anisotropy of Fe, Ni, and Co nanowires
Paulus PM, et al.
Journal of magnetism and magnetic materials, 224(2), 180-196 (2001)
Magnetoresistance of ferromagnetic nanowires
Wegrowe JE, et al.
Physical Review Letters, 82(18), 3681-3684 (1999)
T M Whitney et al.
Science (New York, N.Y.), 261(5126), 1316-1319 (1993-09-03)
Arrays of ferromagnetic nickel and cobalt nanowires have been fabricated by electrochemical deposition of the metals into templates with nanometer-sized pores prepared by nuclear track etching. These systems display distinctive characteristics because of their one-dimensional microstructure. The preferred magnetization direction
High energy product developed from cobalt nanowires.
Gandha K
Scientific Reports, 4 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service