Skip to Content
MilliporeSigma
All Photos(1)

Documents

745510

Sigma-Aldrich

4-Heptyl acetate

98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18O2
Molecular Weight:
158.24
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

Assay

98% (CP)

form

liquid

SMILES string

CCCC(OC(C)=O)CCC

InChI

1S/C9H18O2/c1-4-6-9(7-5-2)11-8(3)10/h9H,4-7H2,1-3H3

InChI key

JPXGPRBLTIYFQG-UHFFFAOYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.4 °F

Flash Point(C)

68 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service