Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

745502

Sigma-Aldrich

4-Heptyl-3-13C acetate

99 atom % 13C, 98% (CP)

Synonym(s):

13C Labeled 4-Heptyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC8H18O2
Molecular Weight:
159.23
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

liquid

mass shift

M+1

SMILES string

CCCC(OC(C)=O)[13CH2]CC

InChI

1S/C9H18O2/c1-4-6-9(7-5-2)11-8(3)10/h9H,4-7H2,1-3H3/i6+1

InChI key

JPXGPRBLTIYFQG-PTQBSOBMSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.4 °F

Flash Point(C)

68 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service