Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

745499

Sigma-Aldrich

Zinc trifluoroethanesulfinate

Synonym(s):

Baran TFES Reagent, Baran Trifluoroethylation Reagent, Bis(((2,2,2-trifluoroethyl)sulfinyl)oxy)zinc, TFES

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4F6O4S2Zn
CAS Number:
Molecular Weight:
359.58
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: C-C Bond Formation
reagent type: catalyst
reaction type: C-H Activation

mp

114-120 °C

functional group

fluoro
sulfinic acid

SMILES string

O=S(O[Zn]OS(CC(F)(F)F)=O)CC(F)(F)F

InChI

1S/2C2H3F3O2S.Zn/c2*3-2(4,5)1-8(6)7;/h2*1H2,(H,6,7);/q;;+2/p-2

InChI key

PUGHSSOALZPRJD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Zinc trifluoroethanesulfinate (TFES) is part of a zinc sulfinate toolbox developed by Phil Baran for the simple and rapid diversification of heterocycles.

Practical and Innate Carbon-Hydrogen Functionalization of Heterocycles

Learn More at the Professor and Product Portal of Professor Phil S. Baran.

Linkage

Frequently Asked Questions are available for this Product.

Other Notes

Previously sold under product number L511234

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuta Fujiwara et al.
Nature, 492(7427), 95-99 (2012-12-04)
Nitrogen-rich heterocyclic compounds have had a profound effect on human health because these chemical motifs are found in a large number of drugs used to combat a broad range of diseases and pathophysiological conditions. Advances in transition-metal-mediated cross-coupling have simplified
Fionn O'Hara et al.
Journal of the American Chemical Society, 135(32), 12122-12134 (2013-07-19)
Radical addition processes can be ideally suited for the direct functionalization of heteroaromatic bases, yet these processes are only sparsely used due to the perception of poor or unreliable control of regiochemistry. A systematic investigation of factors affecting the regiochemistry

Articles

Rapidly diversify (hetero)aromatic scaffolds for chemical industry needs amid resource and time constraints, ensuring efficiency.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service