Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

743046

Sigma-Aldrich

1,3-Bis[(1S)-2,2-dimethyl-1-(1-naphthyl)propyl]imidazolium iodide

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H37IN2
CAS Number:
Molecular Weight:
588.56
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +75±3°, c = 1 in dichloromethane

impurities

≤2.0% water

SMILES string

[I-].CC(C)(C)[C@@H](c1cccc2ccccc12)n3cc[n+](c3)[C@@H](c4cccc5ccccc45)C(C)(C)C

InChI

1S/C33H37N2.HI/c1-32(2,3)30(28-19-11-15-24-13-7-9-17-26(24)28)34-21-22-35(23-34)31(33(4,5)6)29-20-12-16-25-14-8-10-18-27(25)29;/h7-23,30-31H,1-6H3;1H/q+1;/p-1/t30-,31+;

InChI key

DZXSGMCSEFOYEQ-USHPRDJASA-M

Application

Carbene ligand precursor

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masafumi Nakanishi et al.
Chimia, 66(4), 241-243 (2012-05-23)
New sterically hindered chiral N-heterocyclic carbene (NHC) ligands were used in palladium catalysis to bring about a highly enantioselective C(sp(3))-H activation on the methylene site of a cycloalkane moiety. The intramolecular coupling reaction of a prochiral N-aryl-N-cycloalkyl methyl carbamate required
Banerjee, D.; et al.
Organometallics, 31, 709-709 (2012)
Nakanishi, M.; et al.
Angewandte Chemie (International Edition in English), 50, 9775-9775 (2011)
Fused indolines by palladium-catalyzed asymmetric C-C coupling involving an unactivated methylene group.
Masafumi Nakanishi et al.
Angewandte Chemie (International ed. in English), 50(32), 7438-7441 (2011-07-05)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service