Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

741574

Sigma-Aldrich

N,N′-Diisopropyl-13C6-urea

99 atom % 13C, 97% (CP)

Synonym(s):

1,3-Diisopropyl-13C6-urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6CH16N2O
Molecular Weight:
150.17
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

97% (CP)

mass shift

M+6

SMILES string

[13CH3][13CH]([13CH3])NC(=O)N[13CH]([13CH3])[13CH3]

InChI

1S/C7H16N2O/c1-5(2)8-7(10)9-6(3)4/h5-6H,1-4H3,(H2,8,9,10)/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

BGRWYRAHAFMIBJ-IDEBNGHGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service