Skip to Content
MilliporeSigma
All Photos(2)

Documents

741221

Sigma-Aldrich

Bis(benzonitrile)palladium(II) bromide

95%

Synonym(s):

Bis(benzonitrile)dibromopalladium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2(C6H5CN) · PdBr2
CAS Number:
Molecular Weight:
472.47
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

95-102 °C

SMILES string

Br[Pd]Br.N#Cc1ccccc1.N#Cc2ccccc2

InChI

1S/2C7H5N.2BrH.Pd/c2*8-6-7-4-2-1-3-5-7;;;/h2*1-5H;2*1H;/q;;;;+2/p-2

InChI key

OKDHIPXOBPPHTG-UHFFFAOYSA-L

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service