Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

741140

Sigma-Aldrich

4-Chloro-α-cyanocinnamic acid

97% (CP)

Synonym(s):

3-(4-Chlorophenyl)-2-cyano-2-propenoic acid, 3-(4-Chlorophenyl)-2-cyanoacrylic acid, p-Chloro-α-cyanocinnamic acid, p-Chlorobenzylidenecyanoacetic acid, ClCCA, NSC 2479

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6ClNO2
CAS Number:
Molecular Weight:
207.61
Beilstein:
2616773
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

Assay

97% (CP)

form

solid

SMILES string

OC(=O)\C(=C\c1ccc(Cl)cc1)C#N

InChI

1S/C10H6ClNO2/c11-9-3-1-7(2-4-9)5-8(6-12)10(13)14/h1-5H,(H,13,14)/b8-5+

InChI key

MXCRRKYUQNHWLJ-VMPITWQZSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service