Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

740446

Sigma-Aldrich

2-(Trifluoromethyl)pyrimidine-5-carboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3F3N2O2
CAS Number:
Molecular Weight:
192.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

170-175 °C

functional group

carboxylic acid
fluoro

storage temp.

2-8°C

SMILES string

OC(=O)c1cnc(nc1)C(F)(F)F

InChI

1S/C6H3F3N2O2/c7-6(8,9)5-10-1-3(2-11-5)4(12)13/h1-2H,(H,12,13)

InChI key

RGLZXVNOHDUMRJ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nico A Contreras et al.
PLoS pathogens, 15(6), e1007890-e1007890 (2019-06-21)
Cytomegalovirus (CMV) is a ubiquitous herpesvirus infecting most of the world's population. CMV has been rigorously investigated for its impact on lifelong immunity and potential complications arising from lifelong infection. A rigorous adaptive immune response mounts during progression of CMV

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service