Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

739022

Sigma-Aldrich

Bis[2-(di-tert-butylphosphino)ethyl]amine solution

10 wt. % in THF

Synonym(s):

Bis(2-(di-tert-butylphosphino)ethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H45NP2
Molecular Weight:
361.53
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

reagent type: ligand

concentration

10 wt. % in THF

refractive index

n20/D 1.418

density

0.911 g/mL at 25 °C

functional group

phosphine

SMILES string

CC(C)(C)P(CCNCCP(C(C)(C)C)C(C)(C)C)C(C)(C)C

InChI

1S/C20H45NP2/c1-17(2,3)22(18(4,5)6)15-13-21-14-16-23(19(7,8)9)20(10,11)12/h21H,13-16H2,1-12H3

InChI key

MJYKYLNPIGDVEF-UHFFFAOYSA-N

Legal Information

Sold under license from Kanata for research purposes only. WO 2004096735.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service