Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

738913

Sigma-Aldrich

6-Bromo-1,3-benzodioxole-5-carbonitrile

97%

Synonym(s):

2-Bromo-4,5-methylenedioxybenzonitrile, 6-Bromopiperonylonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4BrNO2
CAS Number:
Molecular Weight:
226.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

103-108 °C

SMILES string

Brc1cc2OCOc2cc1C#N

InChI

1S/C8H4BrNO2/c9-6-2-8-7(11-4-12-8)1-5(6)3-10/h1-2H,4H2

InChI key

YJAZLOORQHTWFN-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service