Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

738905

Sigma-Aldrich

11-(Ferrocenyl)undecanethiol

95%

Synonym(s):

11-(Mercaptoundecyl)ferrocene

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$512.00

$512.00


Estimated to ship onApril 21, 2025Details


Request a Bulk Order

Select a Size

Change View
250 MG
$512.00

About This Item

Empirical Formula (Hill Notation):
C21H32FeS
CAS Number:
Molecular Weight:
372.39
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

$512.00


Estimated to ship onApril 21, 2025Details


Request a Bulk Order

Assay

95%

form

solid

mp

36-41 °C

storage temp.

2-8°C

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.SCCCCCCCCCCC[C]2[CH][CH][CH][CH]2

InChI

1S/C16H27S.C5H5.Fe/c17-15-11-7-5-3-1-2-4-6-8-12-16-13-9-10-14-16;1-2-4-5-3-1;/h9-10,13-14,17H,1-8,11-12,15H2;1-5H;

InChI key

ZXKSSFMVUQHJKC-UHFFFAOYSA-N

Application

  • Electrochemical Behavior and Anion Influence: Explores the impact of anions on redox behavior and structure of 11-ferrocenyl-1-undecanethiol monolayers on gold electrodes, using CV and SEIRAS techniques (ScienceDirect, 2020).
  • Electrochemical Characterization: Presents a study on the assembly and electrochemical response of 11-ferrocenyl-1-undecanethiol on gold electrodes, highlighting effects of surface structure (ScienceDirect, 2017).
  • Surface Adsorption Studies: Discusses the adsorption characteristics of ferrocenylated gold nanoparticles and their electrochemical applications (ScienceDirect, 2018).
  • Extended Gate FET Biosensors: Describes the use of 11-(Ferrocenyl)undecanethiol in biosensor applications, highlighting its solubility and application in modified electrode chips (Springer, 2020).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service